Assay Details
| Gene Symbol: TEAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062766.1 | XP_017918255.1 | 9 - 10 | 820 | 61 | |
| XM_018062767.1 | XP_017918256.1 | 8 - 9 | 829 | 61 | ||
| GenBank mRNA | JT703162.1 | - | 13 - 14 | 3285 | 61 | |
| JT746294.1 | - | 10 - 11 | 1800 | 61 | ||
More Information
Set Membership: |
|

TEAD2
Genomic Map