Assay Details
| Gene Symbol: LOC102169177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050674.1 | XP_017906163.1 | 3 - 4 | 1675 | 84 | |
| GenBank mRNA | JT635102.1 | - | 2 - 3 | 596 | 84 | |
More Information
Set Membership: |
|

LOC102169177
Genomic Map