Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967047.2 | XP_013822501.1 | 1 - 2 | 267 | 89 | |
| XM_018054379.1 | XP_017909868.1 | 1 - 2 | 267 | 89 | ||
| GenBank mRNA | KA340729.1 | - | 1 - 2 | 245 | 89 | |
More Information
Set Membership: |
|

LRR1
Genomic Map