Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682091.2 | XP_005682148.1 | 5 - 6 | 590 | 57 | |
| XM_005682092.2 | XP_005682149.1 | 5 - 6 | 587 | 57 | ||
| XM_013964961.2 | XP_013820415.1 | 4 - 5 | 595 | 57 | ||
| XM_013964962.2 | XP_013820416.1 | 4 - 5 | 578 | 57 | ||
| GenBank mRNA | KA312681.1 | - | 4 - 5 | 609 | 57 | |
More Information
Set Membership: |
|

RFXANK
Genomic Map