Assay Details
| Gene Symbol: LOC102183030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040513.1 | XP_017896002.1 | 4 - 5 | 844 | 75 | |
| XM_018040514.1 | XP_017896003.1 | 3 - 4 | 398 | 75 | ||
| XM_018040515.1 | XP_017896004.1 | 4 - 5 | 825 | 75 | ||
More Information
Set Membership: |
|

LOC102183030
Genomic Map