Assay Details
| Gene Symbol: LOC102183030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040513.1 | XP_017896002.1 | 5 - 6 | 946 | 121 | |
| XM_018040514.1 | XP_017896003.1 | 4 - 5 | 500 | 121 | ||
More Information
Set Membership: |
|

LOC102183030
Genomic Map