Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676426.2 | XP_005676483.1 | 3 - 4 | 1326 | 63 | |
| XM_005676429.3 | XP_005676486.1 | 3 - 4 | 1124 | 63 | ||
| XM_018060367.1 | XP_017915856.1 | 3 - 4 | 1329 | 63 | ||
| GenBank mRNA | JT671845.1 | - | 5 - 6 | 1950 | 63 | |
| KU041750.1 | - | 3 - 4 | 915 | 63 | ||
More Information
Set Membership: |
|

KLF7
Genomic Map