Assay Details
| Gene Symbol: PPP1R14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699968.3 | XP_005700025.3 | 2 - 3 | 599 | 73 | |
| GenBank mRNA | JO510384.2 | - | 2 - 3 | 360 | 73 | |
More Information
Set Membership: |
|

PPP1R14B
Genomic Map