Assay Details
| Gene Symbol: CCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696305.3 | XP_005696362.1 | 4 - 5 | 842 | 67 | |
| XM_018038733.1 | XP_017894222.1 | 4 - 5 | 906 | 67 | ||
| GenBank mRNA | JO429302.2 | - | 1 - 2 | 1167 | 67 | |
More Information
Set Membership: |
|

CCND3
Genomic Map