Assay Details
| Gene Symbol: PPP1R1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047639.1 | XP_017903128.1 | 3 - 4 | 316 | 53 | |
| XM_018047640.1 | XP_017903129.1 | 3 - 4 | 315 | 53 | ||
| GenBank mRNA | JO528673.2 | - | 4 - 5 | 362 | 53 | |
More Information
Set Membership: |
|

PPP1R1A
Genomic Map