Assay Details
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051051.1 | XP_017906540.1 | 5 - 6 | 2136 | 64 | |
XM_018051052.1 | XP_017906541.1 | 5 - 6 | 2805 | 64 | ||
GenBank mRNA | JO497518.2 | - | 1 - 2 | 256 | 64 | |
JO497553.2 | - | 1 - 2 | 256 | 64 | ||
JO644759.2 | - | 4 - 5 | 2985 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
