Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687528.3 | XP_005687585.2 | 3 - 4 | 1233 | 65 | |
| XM_013968289.2 | XP_013823743.2 | 2 - 3 | 739 | 65 | ||
More Information
Set Membership: |
|

ZAR1L
Genomic Map