Assay Details
| Gene Symbol: TLE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050799.1 | XP_017906288.1 | 6 - 7 | 495 | 87 | |
| GenBank mRNA | KA328249.1 | - | 1242 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TLE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050799.1 | XP_017906288.1 | 6 - 7 | 495 | 87 | |
| GenBank mRNA | KA328249.1 | - | 1242 | 87 | ||
Set Membership: |
|