Assay Details
| Gene Symbol: NFKBIE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696514.3 | XP_005696571.2 | 3 - 4 | 950 | 57 | |
| XM_018038772.1 | XP_017894261.1 | 3 - 4 | 993 | 57 | ||
| GenBank mRNA | JO598565.2 | - | 3 - 4 | 887 | 57 | |
More Information
Set Membership: |
|

NFKBIE
Genomic Map