Assay Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049322.1 | XP_017904811.1 | 5 - 6 | 1077 | 97 | |
| XM_018049323.1 | XP_017904812.1 | 6 - 7 | 609 | 97 | ||
| XM_018049324.1 | XP_017904813.1 | 4 - 5 | 555 | 97 | ||
| XM_018049325.1 | XP_017904814.1 | 5 - 6 | 1077 | 97 | ||
| XM_018049326.1 | XP_017904815.1 | 5 - 6 | 1077 | 97 | ||
More Information
Set Membership: |
|

LCORL
Genomic Map