Assay Details
| Gene Symbol: YBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042673.1 | XP_017898162.1 | 2 - 3 | 435 | 104 | |
| GenBank mRNA | JO375085.2 | - | 2 - 3 | 178 | 104 | |
| JO635056.2 | - | 2 - 3 | 366 | 104 | ||
More Information
Set Membership: |
|

YBX1
Genomic Map