Assay Details
| Gene Symbol: LOC102189637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038819.1 | XP_017894308.1 | 5 - 6 | 1188 | 75 | |
| XM_018038820.1 | XP_017894309.1 | 5 - 6 | 895 | 75 | ||
| XM_018038821.1 | XP_017894310.1 | 6 - 7 | 969 | 75 | ||
| XM_018038822.1 | XP_017894311.1 | 6 - 7 | 1269 | 75 | ||
| GenBank mRNA | JO426998.1 | - | 2 - 3 | 1864 | 75 | |
More Information
Set Membership: |
|

LOC102189637
Genomic Map