Assay Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968951.2 | XP_013824405.1 | 1 - 2 | 323 | 62 | |
| GenBank mRNA | JO560325.2 | - | 11 - 12 | 1511 | 62 | |
| JO560481.2 | - | 11 - 12 | 1511 | 62 | ||
More Information
Set Membership: |
|

PIGU
Genomic Map