Assay Details
| Gene Symbol: INTS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690788.3 | XP_005690845.1 | 2 - 3 | 255 | 60 | |
| XM_005690789.3 | XP_005690846.1 | 2 - 3 | 255 | 60 | ||
More Information
Set Membership: |
|

INTS11
Genomic Map