Assay Details
| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065610.1 | XP_017921099.1 | 8 - 9 | 962 | 63 | |
| XM_018065611.1 | XP_017921100.1 | 8 - 9 | 1357 | 63 | ||
| GenBank mRNA | KA305375.1 | - | 7 - 8 | 870 | 63 | |
More Information
Set Membership: |
|

TENT4A
Genomic Map