Assay Details
| Gene Symbol: SELENOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709381.3 | XP_005709438.2 | 4 - 5 | 476 | 57 | |
| GenBank mRNA | JO566653.2 | - | 4 - 5 | 298 | 57 | |
| JO632462.2 | - | 2 - 3 | 487 | 57 | ||
More Information
Set Membership: |
|

SELENOW
Genomic Map