Assay Details
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675949.3 | XP_005676006.2 | 9 - 10 | 990 | 94 | |
| XM_018064740.1 | XP_017920229.1 | 9 - 10 | 962 | 94 | ||
| XM_018064757.1 | XP_017920246.1 | 9 - 10 | 883 | 94 | ||
More Information
Set Membership: |
|

MAP3K20
Genomic Map