Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047816.1 | XP_017903305.1 | 9 - 10 | 3472 | 68 | |
| XM_018047817.1 | XP_017903306.1 | 9 - 10 | 3442 | 68 | ||
| XM_018047818.1 | XP_017903307.1 | 8 - 9 | 3352 | 68 | ||
| XM_018047819.1 | XP_017903308.1 | 8 - 9 | 3310 | 68 | ||
| GenBank mRNA | JO620981.2 | - | 1 - 2 | 1720 | 68 | |
More Information
Set Membership: |
|

NCKAP5L
Genomic Map