Assay Details
| Gene Symbol: TRIM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689839.2 | XP_005689896.1 | 3 - 4 | 652 | 64 | |
| XM_005689840.3 | XP_005689897.1 | 3 - 4 | 782 | 64 | ||
| XM_013969741.2 | XP_013825195.1 | 2 - 3 | 193 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013969741.2 transcripts.
Set Membership: |
|

TRIM68
Genomic Map