Assay Details
| Gene Symbol: ZBTB49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049724.1 | XP_017905213.1 | 6 - 7 | 1576 | 90 | |
| XM_018049725.1 | XP_017905214.1 | 6 - 7 | 1581 | 90 | ||
| GenBank mRNA | KA352291.1 | - | 2 - 3 | 1181 | 90 | |
More Information
Set Membership: |
|

ZBTB49
Genomic Map