Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045078.1 | XP_017900567.1 | 7 - 8 | 2251 | 64 | |
| XM_018045079.1 | XP_017900568.1 | 7 - 8 | 2249 | 64 | ||
| XM_018045080.1 | XP_017900569.1 | 7 - 8 | 2249 | 64 | ||
| XM_018045081.1 | XP_017900570.1 | 8 - 9 | 2082 | 64 | ||
| XM_018045083.1 | XP_017900572.1 | 8 - 9 | 2080 | 64 | ||
| GenBank mRNA | JO339061.2 | - | 7 - 8 | 1071 | 64 | |
| JO537611.2 | - | 7 - 8 | 1222 | 64 | ||
More Information
Set Membership: |
|

OTUD5
Genomic Map