Assay Details
| Gene Symbol: FKBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285571.2 | NP_001272500.1 | 6 - 7 | 1026 | 66 | |
| XM_018049966.1 | XP_017905455.1 | 7 - 8 | 1152 | 66 | ||
| XM_018049967.1 | XP_017905456.1 | 7 - 8 | 1220 | 66 | ||
| XM_018049968.1 | XP_017905457.1 | 7 - 8 | 1149 | 66 | ||
| GenBank mRNA | JF714970.1 | - | 6 - 7 | 1029 | 66 | |
| JO580589.1 | - | 2 - 3 | 714 | 66 | ||
More Information
Set Membership: |
|

FKBP8
Genomic Map