Assay Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062128.1 | XP_017917617.1 | 2 - 3 | 1117 | 57 | |
GenBank mRNA | JO642755.2 | - | 1 - 2 | 296 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
