Assay Details
| Gene Symbol: TBC1D25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044879.1 | XP_017900368.1 | 2 - 3 | 389 | 60 | |
| GenBank mRNA | JO401492.2 | - | 4 - 5 | 941 | 60 | |
| KA313205.1 | - | 4 - 5 | 1569 | 60 | ||
More Information
Set Membership: |
|

TBC1D25
Genomic Map