Assay Details
Gene Symbol: LOC102183271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO631943.2 | - | 1509 | 93 |
Gene Symbol: APBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052054.1 | XP_017907543.1 | 2 - 3 | 1461 | 93 | |
XM_018052056.1 | XP_017907545.1 | 2 - 3 | 1356 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
