Assay Details
| Gene Symbol: FUCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684863.3 | XP_005684920.3 | 2 - 3 | 1213 | 65 | |
| GenBank mRNA | JO559609.2 | - | 5 - 6 | 1698 | 65 | |
| JO559626.2 | - | 5 - 6 | 1698 | 65 | ||
More Information
Set Membership: |
|

FUCA2
Genomic Map