Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695598.3 | XP_005695655.2 | 3 - 4 | 833 | 100 | |
| XM_005695599.3 | XP_005695656.2 | 3 - 4 | 790 | 100 | ||
More Information
Set Membership: |
|

ZNF445
Genomic Map