Assay Details
| Gene Symbol: FGF22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683284.3 | XP_005683341.2 | 1 - 2 | 311 | 70 | |
| XM_013965510.2 | XP_013820964.2 | 1 - 2 | 311 | 70 | ||
| GenBank mRNA | JO624544.2 | - | 1 - 2 | 200 | 70 | |
| KA331441.1 | - | 1 - 2 | 293 | 70 | ||
More Information
Set Membership: |
|

FGF22
Genomic Map