Assay Details
| Gene Symbol: VANGL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046377.1 | XP_017901866.1 | 5 - 6 | 1420 | 60 | |
| XM_018046378.1 | XP_017901867.1 | 5 - 6 | 1162 | 60 | ||
More Information
Set Membership: |
|

VANGL2
Genomic Map