Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050545.1 | XP_017906034.1 | 9 - 10 | 1564 | 76 | |
| XM_018050546.1 | XP_017906035.1 | 9 - 10 | 1513 | 76 | ||
| XM_018050547.1 | XP_017906036.1 | 9 - 10 | 1543 | 76 | ||
More Information
Set Membership: |
|

ZNF496
Genomic Map