Assay Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681502.3 | XP_005681559.2 | 4 - 5 | 1228 | 76 | |
| XM_005681503.3 | XP_005681560.2 | 4 - 5 | 1194 | 76 | ||
| GenBank mRNA | JO645188.2 | - | 4 - 5 | 1173 | 76 | |
| KA347071.1 | - | 4 - 5 | 1005 | 76 | ||
| KU041753.1 | - | 3 - 4 | 728 | 76 | ||
More Information
Set Membership: |
|

KLF3
Genomic Map