Assay Details
| Gene Symbol: NAA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054097.1 | XP_017909586.1 | 3 - 4 | 1048 | 90 | |
| XM_018054098.1 | XP_017909587.1 | 3 - 4 | 827 | 90 | ||
| GenBank mRNA | KA339984.1 | - | 2 - 3 | 769 | 90 | |
More Information
Set Membership: |
|

NAA30
Genomic Map