Assay Details
| Gene Symbol: CLVS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684505.3 | XP_005684562.1 | 2 - 3 | 1023 | 68 | |
| XM_018052989.1 | XP_017908478.1 | 2 - 3 | 1203 | 68 | ||
| GenBank mRNA | JO636666.2 | - | 2 - 3 | 1568 | 68 | |
More Information
Set Membership: |
|

CLVS2
Genomic Map