Assay Details
| Gene Symbol: LOC102186637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675805.3 | XP_005675862.2 | 23 - 24 | 3934 | 103 | |
| XM_018066077.1 | XP_017921566.1 | 23 - 24 | 3934 | 103 | ||
More Information
Set Membership: |
|

LOC102186637
Genomic Map