Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065562.1 | XP_017921051.1 | 5 - 6 | 7267 | 62 | |
| XM_018065563.1 | XP_017921052.1 | 4 - 5 | 7390 | 62 | ||
| GenBank mRNA | JO632153.2 | - | 2 - 3 | 1870 | 62 | |
More Information
Set Membership: |
|

MAP1B
Genomic Map