Assay Details
Gene Symbol: RPRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962442.2 | XP_013817896.1 | 8 - 9 | 1923 | 66 | |
XM_018046033.1 | XP_017901522.1 | 9 - 10 | 1994 | 66 | ||
XM_018046034.1 | XP_017901523.1 | 8 - 9 | 1353 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
