Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685736.3 | XP_005685793.1 | 3 - 4 | 580 | 128 | |
| XM_005685737.3 | XP_005685794.1 | 3 - 4 | 525 | 128 | ||
| XM_005685738.3 | XP_005685795.1 | 3 - 4 | 831 | 128 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map