Assay Details
| Gene Symbol: RAB5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048098.1 | XP_017903587.1 | 5 - 6 | 670 | 77 | |
| XM_018048099.1 | XP_017903588.1 | 4 - 5 | 550 | 77 | ||
| XM_018048100.1 | XP_017903589.1 | 4 - 5 | 814 | 77 | ||
| XM_018048101.1 | XP_017903590.1 | 5 - 6 | 685 | 77 | ||
| XM_018048102.1 | XP_017903591.1 | 4 - 5 | 565 | 77 | ||
| XM_018048103.1 | XP_017903592.1 | 3 - 4 | 503 | 77 | ||
| GenBank mRNA | JO633442.2 | - | 1 - 2 | 104 | 77 | |
More Information
Set Membership: |
|

RAB5B
Genomic Map