Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047428.1 | XP_017902917.1 | 5 - 6 | 4942 | 66 | |
| XM_018047435.1 | XP_017902924.1 | 5 - 6 | 1457 | 66 | ||
| XM_018047438.1 | XP_017902927.1 | 5 - 6 | 1436 | 66 | ||
| XM_018047441.1 | XP_017902930.1 | 6 - 7 | 1568 | 66 | ||
More Information
Set Membership: |
|

KLHL24
Genomic Map