Assay Details
| Gene Symbol: LOC102173916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041840.1 | XP_017897329.1 | 2 - 3 | 352 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102173916| Gene Symbol: LOC102173916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041840.1 | XP_017897329.1 | 2 - 3 | 352 | 95 | |
Set Membership: |
|