Assay Details
| Gene Symbol: GPAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049681.1 | XP_017905170.1 | 6 - 7 | 1284 | 67 | |
| GenBank mRNA | JO380617.2 | - | 5 - 6 | 793 | 67 | |
More Information
Set Membership: |
|

GPAT3
Genomic Map