Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 39 - 40 | 6540 | 112 | |
| GenBank mRNA | JO426300.1 | - | 6 - 7 | 928 | 112 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map