Assay Details
| Gene Symbol: LOC102172692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047601.1 | XP_017903090.1 | 5 - 6 | 828 | 68 | |
| Gene Symbol: LOC102187742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO428351.1 | - | 8 - 9 | 1239 | 68 | |
More Information
Set Membership: |
|

LOC102172692
Genomic Map