Assay Details
| Gene Symbol: RBM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688508.3 | XP_005688565.1 | 10 - 11 | 1254 | 88 | |
| XM_005688509.3 | XP_005688566.1 | 10 - 11 | 1259 | 88 | ||
| XM_005688510.3 | XP_005688567.1 | 9 - 10 | 1188 | 88 | ||
| XM_005688511.3 | XP_005688568.1 | 9 - 10 | 1193 | 88 | ||
| XM_018057787.1 | XP_017913276.1 | 11 - 12 | 1327 | 88 | ||
| XM_018057788.1 | XP_017913277.1 | 11 - 12 | 1326 | 88 | ||
| XM_018057789.1 | XP_017913278.1 | 12 - 13 | 1399 | 88 | ||
| XM_018057790.1 | XP_017913279.1 | 10 - 11 | 1221 | 88 | ||
| XM_018057791.1 | XP_017913280.1 | 11 - 12 | 1293 | 88 | ||
| GenBank mRNA | JO419672.2 | - | 7 - 8 | 1589 | 88 | |
| JO419673.2 | - | 7 - 8 | 1571 | 88 | ||
More Information
Set Membership: |
|

RBM39
Genomic Map