Assay Details
| Gene Symbol: FUZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062811.1 | XP_017918300.1 | 2 - 3 | 449 | 59 | |
| GenBank mRNA | JO531114.2 | - | 1 - 2 | 146 | 59 | |
More Information
Set Membership: |
|

FUZ
Genomic Map